Dualmat™ Static Dissipative Mats, Precut, Dark Gray, ACL
Supplier: ACL Staticide
8385DGYM3060 8385DGYM3072 8385DGYM3660 8385DGYM3672 8385DGYM2436 8385DGYM2448 8385DGYM2460 8385DGYM2472
10856-354EA 367.92 CAD
10856-354 10856-356 10856-358 10856-360 10856-368 10856-370 10856-372 10856-374
Dualmat™ Static Dissipative Mats, Precut, Dark Gray, ACL
Surface Protectors Bench Protectors
Dualmat™ is a heavy duty ESD-safe rubber material designed for use on tables and grounded work bench surfaces
These dual layer precut mats feature two 10mm female snaps at the top and bottom left side. Snaps can be located elsewhere if specified. They are available in 8 standard sizes, and custom sizes are also available. Dualmats™ are low outgassing and low VOC, making them ideal for use in cleanrooms and other controlled environments. Additionally, they are UV protected so the color will not fade.
Dissipative on the colored side and conductive on the back side, the design makes itMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable