Single-Stage Ultra-High Purity Chrome-Plated Brass Gas Regulators with International Fittings, Restek
Supplier: Restek
22375 22372 22373 22374 22138
10854-624EA 958.69 CAD
10854-624 10854-626 10854-628 10854-630 10854-748 10854-910
Single-Stage Ultra-High Purity Chrome-Plated Brass Gas Regulators with International Fittings, Restek
Gas Regulators
UHP brass regulators are the best choice when using ultra-high-purity carrier gas for sensitive GC applications using MS, PID, or ECD detection methods.
  • Use when there is secondary pressure regulation downstream
  • Identical gas purity protection as with dual-stage gas regulators
  • Oxidation-resistant
  • Chrome-plated
All regulators are rated to 3000 psig maximum inlet pressure.