Low Pressure Seal for PerkinElmer HPLC Systems, Restek
Supplier: Restek
25435
10853-730EA 139.36 CAD
10853-730
Low Pressure Seal for PerkinElmer HPLC Systems, Restek
Seals Mechanical Seals
These seals made of ultra-high molecular weight polyethylene (UHMWPE); increased resistance to abrasion, best for buffers.