Home Care Bags, Elkay Plastics
Supplier: Elkay Plastics
10G-055415 10G-084021 10G-128030 10G150924 12G-128030 15G-161436 15G-242048 20G-108024 20G-201830 20G-282460 7G042012 7G063012 7G063015 7G084018 8G084018 8G084021 BOR15G-242048T BOR20F2024T BOR2636B BOR4848T BORR161454
10852-194CS 149.09 CAD
10852-194 10852-196 10852-200 10852-202 10852-204 10852-222 10852-224 10852-234 10852-236 10852-238 10852-250 10852-252 10852-254 10852-256 10852-258 10852-260 10852-264 10852-266 10852-268 10852-270 10852-272
Home Care Bags, Elkay Plastics
Bags Storage Bags
Elkay has a full range of DME equipment covers available for all types of accreditation purposes.
  • Made from seamless tubing with no slits or side seals that can rip
  • Feature creased sides that expand to accommodate bulky items
  • Cases and inner packs are bar coded and clearly labeled for easy identification
Low Density Polyethylene (LDPE) bags are the most used plastic bag because they provide excellent protection and outstanding value. These bags are available in Clear, Blue, Tan, Red tints and various sizes. Many of these bags are packed on convenientMore Product Information