Rtx®-CLPesticides Column Kit (0.53 mm ID), Restek
Supplier: Restek
11197
10850-858EA 3012.97 CAD
10850-858
Rtx®-CLPesticides Column Kit (0.53 mm ID), Restek
Chromatography Columns
Improved resolution and faster analyses, compared to 1701 or phenyl phases, make these the pesticide columns of choice.
  • Application-specific columns for organochlorine pesticides and herbicides
  • Low bleed—ideal for high sensitivity GC-ECD or GC-MS analyses
  • Baseline separations in less than 10 minutes.
  • Stable to 320 to 340 °C
  • Analyze EPA method 8081B, 8082A, 8151A, 504.1, 515, 508.1, and 552.2 compounds without time-consuming column changes
Rtx®-CLPesticides columns are specially designed to overcome the coelutions and analyte breakdown typicallyMore Product Information