Rtx®-Dioxin2 Columns (Fused Silica), Restek
Supplier: Restek
10758 10759
10057-828EA 2915.22 CAD
10057-828 10849-742
Rtx®-Dioxin2 Columns (Fused Silica), Restek
Chromatography Columns
Rtx®-Dioxin2 columns are constructed of fused silica and can achieve isomer specificity applications.
  • Isomer specificity for 2,3,7,8-TCDD and 2,3,7,8-TCDF achieved with one GC column
  • Thermally stable to 340 °C for longer lifetime
  • Unique selectivity for toxic dioxin and furan congeners allows use as a confirmation GC column
  • Proprietary Crossbond® phase
These columns are thermally stable and provide a longer lifetime as a result.
Order Now
SPECIFICATIONS
Description 10057-828 10849-742
Environmentally Preferable
Temperature limit 20 to 320/340 °C 20 to 320/340 °C