Rt®-βDEXsp Columns (fused silica), Restek
Supplier: Restek
13110 13111
10848-754EA 2211.88 CAD
10848-754 10849-512
Rt®-βDEXsp Columns (fused silica), Restek
Chromatography Columns
By adding β or γ cyclodextrin to Restek's bonded Rtx®-1701 stationary phase, greatly enhance overall utility and column lifetime for Restek's chiral columns, compared to columns that have pure cyclodextrin stationary phases.
  • 2,3-di-O-propyl-6-O-tert-butyl dimethylsilyl beta cyclodextrin added into 14% cyanopropylphenyl/86% dimethyl polysiloxane
  • Improved column lifetime
  • Enhanced column utility
Separations of more than one hundred chiral compounds have been achieved using Restek's unique DEX columns, and Restek's columns continue to demonstrateMore Product Information