Showa 6110PF Powder-Free Nitrile Gloves, Biodegradable, Showa
Supplier: Showa-Best Glove
6110PFS 6110PFM 6110PFL 6110PFXL 6110PFXXL
10842-380BX 57.84 CAD
10842-380 10842-382 10842-384 10842-386 10842-388 10842-390
Showa 6110PF Powder-Free Nitrile Gloves, Biodegradable, Showa
Gloves
These versatile, biodegradable single-use gloves are highly dexterous and waterproof, whilst upholding extreme comfort during long periods of use.
  • Form-fitting and lightweight
  • Disposable
  • Latex-free, powder-free
  • Water-resistant
  • Textured fingertips for improved grip
Approved for food handling and contact, these gloves are ideal for food preparation, use in laboratories, and general repair or maintenance work.
By using SHOWA’s Eco Best Technology®, the 6110PF’s are completely eco-friendly, breaking down by 82% in just 386 days when tested in aMore Product Information