YMC-Triart Diol-HILIC Hybrid Silica HPLC Columns, Diol-HILIC, 5 µm, Analytical, YMC America
Supplier: YMC America,Inc
TDH12S05-1502WT TDH12S05-1503WT TDH12S05-1546WT TDH12S05-2546WT TDH12S05-0202WT TDH12S05-0302WT TDH12S05-0502WT TDH12S05-0503WT TDH12S05-0546WT TDH12S05-1002WT TDH12S05-1003WT TDH12S05-1046WT TDH12S05-1504WT TDH12S05-1506WT TDH12S05-2504WT TDH12S05-H546WT TDH12S05-L502WT TDH12S05-L503WT TDH12S05-L546WT TDH12S05-R503WT TDH12S05-R504WT TDH12S05-1546PTP TDH12S05-05Q1PTP TDH12S05-10Q1PTP TDH12S05-15Q1PTP TDH12S05-0546PTP TDH12S05-1046PTP
89238-170EA 1982.77 CAD
89238-170 89238-172 89238-174 89238-176 10842-168 10842-176 10842-178 10842-180 10842-182 10842-184 10842-186 10842-188 10842-190 10842-192 10842-194 10842-196 10842-198 10842-200 10842-202 10842-204 10842-206 75995-058 75995-226 75995-228 75995-230 75995-232 75995-234
YMC-Triart Diol-HILIC Hybrid Silica HPLC Columns, Diol-HILIC, 5 µm, Analytical, YMC America
Chromatography Columns
Triart Diol-HILIC is a dihydroxypropane moiety bonded to an organic/inorganic hybrid based silica particle.
  • Dihydroxypropyl phase bonded to organic/inorganic hybrid silica particle
  • Ideal for the separation of polar and hydrophilic compounds that are not retained by reversed phase chromatography
  • Superior chemical durability
  • Identical selectivity over particle sizes
  • Ideal for HPLC
YMC-Triart Diol HILIC can retain extremely polar compounds while using eluents and buffers suitable for LC-MS.
The resulting product has excellent durability and usabilityMore Product Information