YMC-Triart C18 Hybrid Silica UHPLC Columns, C18, 1.9 µm, YMC America
Supplier: YMC America,Inc
TA12SP9-0202PT TA12SP9-0302PT TA12SP9-0303PT TA12SP9-0502PT TA12SP9-0503PT TA12SP9-1002PT TA12SP9-1003PT TA12SP9-1502PT TA12SP9-L502PT TA12SP9-L503PT TA12SP9-0546PT TA12SP9-05Q1PT TA12SP9-10Q1PT TA12SP9-1503PT TA12SP9-05Q1PTP TA12SP9-10Q1PTP TA12SP9-15Q1PTP TA12SP9-03Q1PT TA12SP9-15Q1PT TA12SP9-L5Q1PT
89209-866EA 2459.4 CAD
89209-866 89209-868 89209-870 89209-872 89209-874 89209-876 89209-878 89209-880 89209-882 89209-884 10842-052 10842-054 10842-056 10842-058 75994-866 75995-002 75995-004 76012-974 76013-876 76013-878
YMC-Triart C18 Hybrid Silica UHPLC Columns, C18, 1.9 µm, YMC America
Chromatography Columns
YMC-Triart C18, 1.9 µm is a novel, organic/inorganic hybrid silica particle derivatized with C18 (L1 USP designation).
  • Chemical stability of the material combines with inherent mechanical durability of particles to contribute to long column lifetime
  • Available in columns packed with 1.9 µm particles, operating over a wide pH range of 1 to 12; this enables the manipulation of analytical retention and preparative loading
  • Proprietary endcapping makes for excellent peak shapes of all types of compounds
  • Rigorous manufacturing controls result in legendary YMC
More Product Information