Ultra Biphenyl HPLC Columns (USP L11), Restek
Supplier: Restek
9109552 910931E 910933E 910936E 910956E 910957E 9109312 9109315 9109332 9109335 9109352 9109355 9109362 9109365 9109515 9109555 9109562 9109565 9109572 9109575 9109512 910935E 9109362-700 910951E 910952E 910953E 9109552-700 910955E 9109575-700 9109522 9109525 9109532 9109535
97040-724EA 1325.53 CAD
97040-724 10058-354 10058-356 10058-358 10058-360 10058-362 10059-702 10059-704 10059-706 10059-708 10059-712 10059-714 10059-716 10059-718 10059-720 10059-726 10059-728 10059-730 10059-732 10059-734 10803-442 10838-124 10838-126 10838-128 10838-130 10838-134 10838-136 10838-138 10838-140 10838-142 10838-144 10838-146 10838-148 10838-150 10838-152 10838-156 10838-158 10838-160 10838-162 10838-164 10838-166 10838-168 10838-170 10838-172 10838-174 10838-176 10838-178 10839-286 10839-288 10839-292 10839-294 10840-586 10840-588 10840-590 10840-592 10840-596 10840-598
Ultra Biphenyl HPLC Columns (USP L11), Restek
Chromatography Columns
The Ultra line represents Restek’s broadest selection of stationary phases on a single silica support.
  • High-purity type-B silica
  • High-density bonding
  • Designed for selectivity
Since 2005, the Restek biphenyl has offered a greater degree of dispersion than conventional phenyls and a greater degree of polarizability than phenyl hexyls, creating higher selectivity and a greater range of usability. Because of these heightened interactions, this column shows substantial increases in retention—especially for dipolar, unsaturated, or conjugated solutes—andMore Product Information