SP Bel-Art Safety Goggles, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H24873-0000
10837-120EA 70.36 CAD
10837-120
SP Bel-Art Safety Goggles, Bel-Art Products, a part of SP
Goggles
Made of soft, clear, flexible vinyl that conforms to the face and nose, these goggles have four vents to reduce fogging.
  • Soft, clear, flexible vinyl conforms to the face and nose, with four vents to reduce fogging
  • Clear polycarbonate lenses can be worn over regular eyeglasses with complete comfort
  • 13 mm (½") wide adjustable elastic headband
  • ANSI Standard Z87.1 compliant
Goggles have clear polycarbonate lenses, and can be worn over regular eyeglasses with complete comfort. 13 mm (½") wide adjustable elastic headband. Goggles are ANSI Standard Z87.1 compliant.
Order Now
SPECIFICATIONS
Environmentally Preferable