Bench Top AC Ionizer with Heater, Transforming Technologies
Supplier: Transforming Technologies
BFN8412
10837-092EA 2613.1 CAD
10837-092
Bench Top AC Ionizer with Heater, Transforming Technologies
Ionizers
The BFN8412 extended range ionizing blower, with AC technology, provides excellent balance and stability for secure workstation protection
BFN series AC ionizers from Transforming Technologies create a dense and well-balanced ionization current. They are unique in their ability to deliver fast decay times with low offset voltages. Continuous balance and decay protection is assured by the reliable AC design. An integrated heater, emitter point cleaner, removable front and rear fan guards, safety switch combine to make the BFN 8412 a powerful and reliable ionizer.
Designed to cover aMore Product Information