Ionizing Nozzles, Transforming Technologies
Supplier: Transforming Technologies
N0020 N0030 N0040 N0050-12 N0050-16 N0050-20 N0050-8 N0010
10837-062EA 97 CAD
10837-062 10837-064 10837-066 10837-068 10837-070 10837-072 10837-074 10837-076 10837-078 10837-134
Ionizing Nozzles, Transforming Technologies
Ionizers
Transforming Technologies’ Ionizing Nozzles can handle unique and difficult point-of-use ionization applications with interchangeable nozzle tips
Ionizing Nozzles are compatible with all IN Gun series ionizers.
Ionizing nozzles can be mounted in hard to reach areas that larger ionizers simply cannot handle. Nozzle tips are designed to spread ionized air in many unique ways. Wide angle, long range, bendable, luer lock, or manifold tips may be substituted for the N0010 tip, depending upon the application. (N0010 is the standard tip sold with Transforming Technologies Ionizing Nozzles).
Order Now
SPECIFICATIONS
Environmentally Preferable