Ranger™ Series Dual Wire Resistance Monitor, Transforming Technologies
Supplier: Transforming Technologies
CM1600 CM1601 CM1602
10837-010EA 600.29 CAD
10837-010 10837-012 10837-014
Ranger™ Series Dual Wire Resistance Monitor, Transforming Technologies
Static Control Monitors
The Resistance Ranger™ Series is the most reliable workstation monitoring system available
Resistance based monitoring is the most complete wrist strap monitoring system available. Workstations using resistance monitors are almost never at risk for a failed ground connection. This type of monitor is used with a two wire (dual conductor) wrist strap. When a person is wearing a wrist strap, the monitor observes the resistance of the loop, consisting of a wire, a person, a wristband, and a second wire. If any part of the loop should open (become disconnected or have out of limit resistance),More Product Information