Double Filtered Balance Enclosures, Labconco®
Supplier: Labconco
3971280 3971281 3971282 3971380 3971381 3971482 3971485 3971486 3971487 3971490 3971491 3971492 3971585 3971586 3971687 3971690 3971691 3971692 3971591 3971592 3971685 3971382 3971480 3971481 3971587 3971590 3971686
10830-828EA 21464.3 CAD
10830-828 10830-830 10830-832 10830-834 10830-836 10830-838 10830-840 10830-844 10830-846 10830-848 10830-850 10830-852 10830-854 10830-856 10830-858 10830-860 10830-862 10830-866 10830-868 10830-870 10830-872 10830-880 10830-882 10830-884 10830-886 10830-888 10830-890
Double Filtered Balance Enclosures, Labconco®
Fume Hoods and Enclosures Balances Enclosures
The patented RXPert Double Filtered Balance System is a Class I Enclosure that meets the USP <800> requirements for non-sterile compounding with NO DUCTWORK! It provides user protection by keeping powders and particulates contained during hazardous drug manipulation
The RXPert features in intrinsically safe negative pressure design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. This balance enclosure includes 2 HEPA filters to provide redundant HEPA filtration in series. The true bag-in/bag-out HEPA filterMore Product Information