RXPert™ Single Filtered Balance Systems, Labconco®
Supplier: Labconco
3971200 3971201 3971202 3971300 3971301 3971302 3971400 3971401 3971402 3971475 3971476 3971477 3971495 3971496 3971497 3971575 3971576 3971577 3971595 3971596 3971597 3971675 3971676 3971677 3971695 3971696 3971697
10216-844EA 14962.6 CAD
10216-844 10216-846 10216-848 10216-850 10216-852 10216-854 10216-856 10216-858 10216-860 10830-786 10830-788 10830-790 10830-792 10830-800 10830-802 10830-804 10830-806 10830-808 10830-810 10830-812 10830-814 10830-816 10830-818 10830-820 10830-822 10830-824 10830-826
RXPert™ Single Filtered Balance Systems, Labconco®
Fume Hoods and Enclosures Balances Enclosures
The patented RXPert Single Filtered Balance System is a Class I Enclosure that meets the USP<800> requirements for non-sterile compounding
The RXPert features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. This balance enclosure includes an all-in-one factory-installed canopy, and damper allows for thimble ducting to the outside. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement.
The HEPA filter is easily accessed to allow the entire surface to beMore Product Information