RXPert™ Single Filtered Balance Systems, Labconco®
Supplier: Labconco
3971200 3971201 3971202 3971300 3971301 3971302 3971400 3971401 3971402 3971475 3971476 3971477 3971495 3971496 3971497 3971575 3971576 3971577 3971595 3971596 3971597 3971675 3971676 3971677 3971695 3971696 3971697
10216-844EA 14962.6 CAD
10216-844 10216-846 10216-848 10216-850 10216-852 10216-854 10216-856 10216-858 10216-860 10830-786 10830-788 10830-790 10830-792 10830-800 10830-802 10830-804 10830-806 10830-808 10830-810 10830-812 10830-814 10830-816 10830-818 10830-820 10830-822 10830-824 10830-826
RXPert™ Single Filtered Balance Systems, Labconco®
Fume Hoods and Enclosures Balances Enclosures
The patented RXPert Single Filtered Balance System is a Class I Enclosure that meets the USP<800> requirements for non-sterile compounding
The RXPert features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. This balance enclosure includes an all-in-one factory-installed canopy, and damper allows for thimble ducting to the outside. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement.
The HEPA filter is easily accessed to allow the entire surface to beMore Product Information
Order Now
SPECIFICATIONS
Electrical 115V, 60Hz
Environmentally Preferable Sustainable Packaging,Sustainable Materials