VWR® Benchtop General Purpose Centrifuges
Supplier: VWR International
10830-744 10830-746 10830-762 10830-764 10830-766 10830-768 10830-770 10830-772 10830-774 10830-776
10830-744EA 10812.34 CAD
10830-744 10830-746 10830-762 10830-764 10830-766 10830-768 10830-770 10830-772 10830-774 10830-776
VWR® Benchtop General Purpose Centrifuges
Centrifuges Benchtop Centrifuges
This compact benchtop unit offers laboratories a true workflow benefit due to its vast capabilities as mid-sized, general purpose, tabletop centrifuge.
  • Offers a unique fast-cool function
  • General-purpose use
  • Mid-sized tabletop centrifuge is compatible with most laboratory workflows
Available with temperature control, the refrigerated unit offers a unique fast-cool function that will lower the unit to +4 °C in 10-15 minutes saving you valuable time spinning delicate samples.
Whether you are working with blood, urine, cell/tissue culture, plates orMore Product Information