Avantor Sterile Polycarbonate Single-Use Flask Assemblies, Avantor Fluid Handling
Supplier: VWR International
5000012 5000013 5000014 5000010 5000011
10830-278CS 5280.68 CAD
10830-278 10830-280 10830-290 10830-700 10830-702
Avantor Sterile Polycarbonate Single-Use Flask Assemblies, Avantor Fluid Handling
Flasks Cell Culture Flasks
Sterile polycarbonate flasks with weldable tubing provide the maximum amount of flexibility and reliability.
  • Sterile flasks come individually bagged and process ready
  • Multiple flask sizes allow for process-specific flexibility
  • Gamma irradiated to Sterility Assurance Level (SAL) 10-6
These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials
When used with sterile Avantor tube sets, custom possibilities are endless.
Assemblies have been designed with a broad spectrum of applications in mind, includingMore Product Information