5049 Series ESD Jacket/Lab Coat, Transforming Technologies
Supplier: Transforming Technologies
JWC5404LB JWC5404WH JWC5403LB JWC5403WH JWC5402LB JWC5402WH JWC5407SPWH JWC5408SPLB JWC5408SPWH JWC5409SPLB JWC5409SPWH JWC5404SPLB JWC5404SPWH JWC5403SPLB JWC5403SPWH JWC5402SPWH JWC5405SPLB JWC5405SPWH JLC5406SPLB JLC5406SPWH JLC5407SPLB JLC5407SPWH JWC5405LB JWC5405WH JWC5406SPLB JWC5406SPWH JWC5407SPLB JWC5402SPLB JLC5402SPLB JLC5402SPWH JLC5405SPLB JLC5405SPWH JWC5406LB JWC5406WH JWC5407LB JWC5407WH JWC5408LB JWC5408WH JWC5409LB JWC5409WH JLC5408SPLB JLC5408SPWH JLC5409SPLB JLC5409SPWH JLC5404SPLB JLC5404SPWH JLC5403SPLB JLC5403SPWH
10828-598EA 136.48 CAD
10828-598 10828-626 10828-680 10828-682 10828-730 10828-732 10828-812 10828-820 10828-828 10828-836 10828-844 10828-868 10828-876 10828-900 10828-908 10828-916 10828-940 10828-948 10829-058 10829-060 10829-064 10829-066 10829-074 10829-076 10829-084 10829-092 10829-100 10829-108 10829-110 10829-112 10829-116 10829-118 10829-144 10829-146 10829-150 10829-152 10829-156 10829-158 10829-162 10829-164 10829-172 10829-174 10829-178 10829-180 10829-184 10829-186 10829-190 10829-192
5049 Series ESD Jacket/Lab Coat, Transforming Technologies
Workwear
The 5049 series ESD jackets are made with more cotton than common polyester ESD jackets which makes the jackets more comfortable, breathable and softer. 5049 Fabric is made with 66% Polyester, 32% Cotton and 2% Carbon.
  • Garment maintains consistent continuity readings for up to 100 wash cycles
  • Breathable cotton poly fabric blend allows for more airflow, and is an excellent choice for very warm climates
  • Reliable panel-to-panel continuity which eliminates the danger of isolated conductors
  • Available in different lengths
  • Available with different cuff options (
More Product Information