9010 Series ESD Jacket, Transforming Technologies
Supplier: Transforming Technologies
JKC9027SPMR JKC9027SPTL JKC9027SPWH JKC9028SPBK JKC9028SPLB JKC9028SPMR JKC9028SPTL JKC9028SPWH JKC9029SPBK JKC9029SPLB JKC9029SPMR JKC9029SPTL JKC9029SPWH JKC9024SPBK JKC9024SPLB JKC9024SPMR JKC9024SPTL JKC9024SPWH JKC9023SPBK JKC9023SPLB JKC9023SPMR JKC9023SPTL JKC9023SPWH JKC9022SPMR JKC9022SPTL JKC9022SPWH JKC9025SPBK JKC9025SPLB JKC9025SPMR JKC9025SPTL JKC9025SPWH JKC9021SPBK JKC9021SPLB JKC9021SPMR JKC9021SPTL JKC9021SPWH JKC9026SPBK JKC9026SPLB JKC9026SPMR JKC9026SPTL JKC9026SPWH JKC9027SPBK JKC9027SPLB JKC9022SPBK JKC9022SPLB
10828-748EA 125.31 CAD
10828-748 10828-808 10828-810 10828-814 10828-818 10828-822 10828-824 10828-826 10828-830 10828-834 10828-838 10828-840 10828-842 10828-862 10828-866 10828-870 10828-872 10828-874 10828-894 10828-898 10828-902 10828-904 10828-906 10828-910 10828-912 10828-914 10828-934 10828-938 10828-942 10828-944 10828-946 10828-950 10828-952 10828-954 10828-956 10828-958 10829-078 10829-082 10829-086 10829-088 10829-090 10829-094 10829-098 10829-102 10829-106
9010 Series ESD Jacket, Transforming Technologies
Workwear
The 9010 Series ESD Jackets provide high quality ESD protection at a great value
The 9010 Series jackets are fabricated from lightweight 90% polyester and 10% carbon fabric for unmatched operator comfort. Jackets feature a snap front closure, wrist-adjustment snaps, three roomy patch pockets, and lapel style collars. These garment maintains consistent continuity readings for up to 100 wash cycles. Available in blue, black, white, teal, and maroon.
Order Now
SPECIFICATIONS
Model 9010
Environmentally Preferable
Garment application Laboratory
Risk area ESD
Gender Unisex
Sleeve or leg length Long
Closure type Buttons
Fabric 90% polyester and 10% carbon