Human Recombinant IL-6 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-10-100UG 100-10-1MG 100-10-20UG 100-10-5UG 100-10-500UG
10787-140EA 818.52 CAD
10787-140 10787-142 10787-144 10787-146 10822-112
Human Recombinant IL-6 (from E. coli)
Proteins and Peptides
Interleukin 6 (IL-6) is an important pro-inflammatory and anti-inflammatory cytokine expressed by T cells, macrophages, and muscle cells. IL-6 signals through a receptor complex containing two receptors, IL-6Rα and gp130.
  • High quality
  • Low endotoxin
  • Affordable
IL-6 has an important function in promoting fever and can serve to stimulate an immune response to trauma. IL-6 is often used for growth of hybridoma cell lines.  Human IL-6 is active on mouse and rat cells.
Caution: For research use only.