Rat Recombinant GDNF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
300-10-100UG 300-10-10UG 300-10-1MG 300-10-500UG
10788-430EA 1368.83 CAD
10788-430 10788-432 10788-434 10788-436 10822-018
Rat Recombinant GDNF (from E. coli)
Proteins and Peptides
Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic factor that signals through a multicomponent receptor system to activate receptor tyrosine kinase RET signaling. GDNF promotes dopamine uptake, prevents motor neuron apoptosis, and supports the survival and differentiation of neurons.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.