VWR® Pre-Certified Inspection Grade 8" Test Sieves, Brass and Stainless Steel
Supplier: VWR International
10819-506 10819-508 10819-510 10819-512 10819-514 10819-516 10819-518 10819-520 10819-522 10820-124 10820-132 10820-136 10820-138 10820-140 10820-142 10820-144 10820-146 10820-148 10820-150 10820-152 10820-154 10820-156 10820-158 10820-162 10820-166 10820-168 10820-170 10820-172 10820-174 10820-180
10819-506EA 424.77 CAD
10819-506 10819-508 10819-510 10819-512 10819-514 10819-516 10819-518 10819-520 10819-522 10820-124 10820-132 10820-136 10820-138 10820-140 10820-142 10820-144 10820-146 10820-148 10820-150 10820-152 10820-154 10820-156 10820-158 10820-162 10820-166 10820-168 10820-170 10820-172 10820-174 10820-180
VWR® Pre-Certified Inspection Grade 8" Test Sieves, Brass and Stainless Steel
Sieves Test Sieves
Because accuracy and precision are vital components of any Quality Control Program, VWR® Pre-Certified Sieves ensure compliance with the American Standards Testing Materials (ASTM) E11 specifications.
  • 1" Half height or 2" Full Height
  • ASTM E11 Inspection Grade Certification (99.0% Confidence Level)
VWR® Pre-Certified Sieves employ sophisticated software and optical technology that scans for irregularities and takes physical measurements in a midpoint orientation of the wire in the “X” and “Y” directions (warp and weft)
The current ASTM E11 standardMore Product Information