VWR® Pre-Certified 8" Test Sieves, All Stainless Steel
Supplier: VWR International
10820-070 10820-072 10820-074 10820-076 10820-078 10820-080 10820-082 10820-086 10820-088 10820-090 10820-092 10820-094 10820-108 10820-122 10820-274 10820-276 10820-278 10820-282 10820-284 10820-286 10820-288 10820-290 10820-292 10820-294 10820-296 10820-298 10820-300 10820-302 10820-304 10820-306 10820-308 10820-312 10820-314 10820-316 10820-318 10820-320 10820-324
10820-070EA 583.17 CAD
10820-070 10820-072 10820-074 10820-076 10820-078 10820-080 10820-082 10820-086 10820-088 10820-090 10820-092 10820-094 10820-108 10820-122 10820-274 10820-276 10820-278 10820-282 10820-284 10820-286 10820-288 10820-290 10820-292 10820-294 10820-296 10820-298 10820-300 10820-302 10820-304 10820-306 10820-308 10820-312 10820-314 10820-316 10820-318 10820-320 10820-324
VWR® Pre-Certified 8" Test Sieves, All Stainless Steel
Sieves Test Sieves
As with other precision lab instruments, periodic re-certification is recommended to ensure the continued conformance and accuracy of working test sieve.
  • 1" Half height or 2" Full Height
  • ASTM E11 Inspection Grade Certification (99.0% Confidence Level)
Working sieves that are used regularly will eventually suffer wear that may produce inaccurate test results
The current ASTM E11 standard offers two available grades of pre-certified sieves (inspection or calibration). Inspection Grade sieves are supplied with a Sieve Inspection Certificate containingMore Product Information