Flame Resistant Long Sleeve Henley Shirt with Max Comfort™ Material, Gray, MCR Safety
Supplier: MCR Safety
H1GL H1GM H1GS H1GX2 H1GX3 H1GX4 H1GX5 H1GXL
10813-454EA 208.56 CAD
10813-454 10813-456 10813-458 10813-460 10813-462 10813-464 10813-466 10813-468
Flame Resistant Long Sleeve Henley Shirt with Max Comfort™ Material, Gray, MCR Safety
Workwear
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in MCR Safety's Flame Resistant (FR) Gear made with Max Comfort™ fabrics
  • 100% FR Cotton
  • Left chest pocket
  • Moisture wicking and breathable
  • Rib-knit collar and cuff
  • NFPA 2112, NFPA 70E, UL Certified
The H1 Henley Shirt features a 100% FR cotton material for maximum comfort. The Max Comfort™ material meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Garment application Mechanical protection,Industrial,Fire protection
Risk area Anti-static,Heat,Flame-retardant
Gender Unisex
Sleeve or leg length Long
Closure type Buttons
Fabric 100% FR cotton
Thickness 7 oz.