VWR® Air Jacketed CO₂ Incubators
Supplier: VWR International
10810-902 10810-944 10811-002 10811-004
10810-902EA 11123.04 CAD
10810-902 10810-944 10811-002 10811-004
VWR® Air Jacketed CO₂ Incubators
Incubators CO₂ Incubators
Enhanced safety features include HEPA filtration and high heat sterilization cycle. The VWR® Air Jacketed CO2 Incubator offers two levels of security for your valuable cell cultures.
  • 6.5 cu. ft. internal volume
  • Removable water pan and four shelves included
  • In-chamber HEPA filtration airflow designed to protect samples from airborne contaminants during routine access
  • Overnight 140 °C sterilization cycle for easy cleaning and reliable decontamination
  • In-chamber probes and sensors for precise control
  • Choice of long lasting TC or independent IR CO2 sensor
  • Easy
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Electrical 120V