Brady® Chemical, Biohazard, and Hazardous Material Signs: Non-Potable Water, Brady Worldwide
Supplier: Brady Worldwide
84467 84468 84313 84314 69407 22359 22360 69408 40847 40846 22283 22282 40923 40924
10810-344EA 24.63 CAD
10810-344 10810-346 10810-374 10810-376 10810-456 10810-458 10810-460 10810-592 10810-602 10810-604 10810-654 10810-656 10810-678 10810-680
Brady® Chemical, Biohazard, and Hazardous Material Signs: Non-Potable Water, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-
More Product Information