Hazardous Material D.O.T. Vehicle Placards: Explosive, Brady
Supplier: Brady Worldwide
63300 63301 63302 63303 63304 63312 63313 63314 63316 63317 63320 63321 63322 63323 63324 63326 63327 63328 63331 63308 63318 63325 63332 63333 63334 63306 63307 63310 63311
10810-286EA 30.32 CAD
10810-286 10810-288 10810-290 10810-292 10810-294 10810-296 10810-298 10810-300 10810-304 10810-306 10810-308 10810-310 10810-312 10810-314 10810-316 10810-318 10810-320 10810-324 10810-326 10810-328 10810-330 10810-332 10810-334 10810-336 10810-338 10810-502 10810-504 10810-508 10810-510
Hazardous Material D.O.T. Vehicle Placards: Explosive, Brady
Labels
D.O.T
  • Placards meet D.O.T. requirements 49CFR Part 172
  • Self sticking vehicle placards may be applied directly to a vehicle
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil, and water resistance
B-946 vinyl (SSV) placards may be applied directly to a vehicle, and will provide over five years of outdoor durability. All placards come with square corners and no holes. Custom placards are also available upon request.
Vehicle placards help ensure that hazardous material shipmentsMore Product Information