DuPont™ Tyvek® IsoClean® Cleanroom Coveralls
Supplier: DuPont
IC255BWH2X00250B IC255BWH3X00250B IC255BWH4X00250B IC255BWH5X00250B IC255BWH6X00250B IC255BWH7X00250B IC255BWHSM0025CS IC255BWHMD0025CS IC255BWHMD00250B IC255BWHLG00250B IC255BWHXL00250B IC255BWHLG0025CS IC255BWHXL0025CS IC255BWH2X0025CS IC255BWH3X0025CS IC255BWH4X0025CS IC255BWH5X0025CS IC255BWH6X0025CS
10809-042CS 821.99 CAD
10809-042 10809-044 10809-046 10809-112 10809-114 10809-116 10809-118 10809-120 10809-122 10809-124 10809-126 10809-128 10809-138 10809-140 10809-142 10809-144 10809-146 10809-148 10809-150
DuPont™ Tyvek® IsoClean® Cleanroom Coveralls
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Bound seams
  • Elastic thumb loops
  • Bound neck
  • Dolman sleeve design
  • Covered elastic wrists and ankles
  • Zipper closure
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Coverall has elastic openings for tighter fit at wrist and ankle. Bound neck ensures lowerMore Product Information