ESD Inspection Gloves, Transforming Technologies
Supplier: Transforming Technologies
GL4501 GL4502 GL4503 GL4504 GL4505 GL4506 GL4501T GL4502T GL4503T GL4504T GL4505T GL4506T GL4501P GL4502P GL4503P GL4504P GL4505P GL4506P
10808-992CS 89.44 CAD
10808-992 10808-994 10808-996 10808-998 10809-000 10809-002 10809-004 10809-006 10809-008 10809-010 10809-012 10809-014 10809-016 10809-018 10809-020 10809-022 10809-024 10809-026
ESD Inspection Gloves, Transforming Technologies
Gloves
Transforming Technologies ESD inspection gloves are designed to be worn when handling ESD susceptible items.
  • Breathable
  • Laundered for repeated use
Gloves are manufactured from a nylon and conductive carbon fabric, and feature finished rolled cuffs. Tip or palm coatings consist of micro-foamed polyurethane coating.
They are made with a dissipative nylon to reduce charge generation. A seamless fine gauge knit construction makes it the most breathable glove available. Available in plain, polyurethane finger tip coated, or polyurethane palm coatedMore Product Information