VWR® Numbered Animal Ear Tags
Supplier: VWR International
10806-296
10806-296EA 65.42 CAD
10806-296
VWR® Numbered Animal Ear Tags
Animal Supplies Animal ID Tags and Punches
Ear tags are made from an ASTM standard (B209-10) aluminum sheet.
  • Aluminum sheet construction
  • Easy to insert
  • Numbered for quick identification
The small aluminum tags are numbered 1 to 100, and are supplied with 100 washers. To apply, simply insert tag through hole and place washer behind hole. Bend tag ends back in rivet fashion, leaving space for ear growth.