VWR® Adson Bone Rongeur
Supplier: VWR International
10806-254 10806-256
10806-254EA 545.99 CAD
10806-254 10806-256
VWR® Adson Bone Rongeur
Bone Cutters and Rongeurs
The VWR® Adson Bone Rongeur is made from ASTM (899-94) Series 400 german stainless steel.
  • Passivated and surgical grade
  • Rockwell Hardness (HRC): 50–55
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Available in a straight or cuved format, the instrument is designed for cutting cartilage and bone. A strong, removable bar-spring makes these shears self-opening.