Precision™ Shaking Water Baths, Thermo Scientific
Supplier: Thermo Fisher Scientific
TSSWB27 TSSWB15S TSDUB15
10805-308EA 5649.2 CAD
10805-308 10805-310 10805-312 10805-314
Precision™ Shaking Water Baths, Thermo Scientific
Baths Shaking Water Baths
Thermo Scientific™ Precision™ shaking water baths support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing
Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere.
Certifications: cULus Listed, CE compliant, US FDA Class I MedicalMore Product Information