VWR® Specimen Containers with Dual Click
Supplier: VWR International
10803-950 10803-952 10803-956 10803-960 10803-962 10803-964 10803-968 10803-972 10803-974 10803-976 10803-978 10803-984 10803-986 10803-988 10803-990 10803-992 10803-994 10803-996 10803-998 10804-000 10804-004 10804-012 10804-018 10804-024 10804-034 10804-036 10804-042 10804-046 10804-048 10804-050 10804-052
10803-950CS 628.5 CAD
10803-950 10803-952 10803-956 10803-960 10803-962 10803-964 10803-968 10803-972 10803-974 10803-976 10803-978 10803-984 10803-986 10803-988 10803-990 10803-992 10803-994 10803-996 10803-998 10804-000 10804-004 10804-012 10804-018 10804-024 10804-034 10804-036 10804-042 10804-046 10804-048 10804-050 10804-052
VWR® Specimen Containers with Dual Click
Sample Containers General Purpose Containers
The VWR® Dual Click line of specimen containers are manufactured from virgin medical grade propylene, and the Dual Click design features two audible and tactile “clicks” to let the user know the lid is properly secured.
  • Virgin medical grade propylene construction
  • Audible and tactile clicks ensure a secure seal
  • Available in sterile and nonsterile formats
Dual Click closures ensure safe specimen transport
Sterile Dual Click specimen containers are sterilized using E-Beam sterilization. Individually wrapped sterile Dual Click containers are alsoMore Product Information