HPLC 30 - Column Storage Cabinet, Restek
Supplier: Restek
25159
10803-376EA 729.06 CAD
10803-376
HPLC 30 - Column Storage Cabinet, Restek
Furniture Storage Cabinets
This easy-to-install cabinet saves space and protects columns.
  • Saves space and protects columns
  • Hinged door is clear for quick identification of column labels or tags
  • Made from solid, durable PVC
  • Easy-to-install, wall-mountable (screws not included), securely holds 30 columns (minimum 100 mm in length)
The hinged door is clear to allow quick identification of column labels or tags.
Ordering information: Columns in photograph are not included.