VWR® One-Unit Counter
Supplier: VWR International
10799-196
10799-196EA 178.62 CAD
10799-196
VWR® One-Unit Counter
Cash and Check Handling Supplies Tally Counters
These compact chromed metal counters allow for quick and easy counting one digit at a time.
  • Compact device tallies up to 9999
  • Chrome-finished metal construction for reliability, long life, and hard usage
  • White numerals on black background for ease in reading
  • Thumb pressure on button produces an audible sound