VWR® Disposable Petri Dishes
Supplier: VWR International
25384-062 25384-088 25384-090 25384-092 25384-093 25384-164 25384-166 25384-168 25384-302 25384-304 25384-306 25384-308 25384-310 25384-312 25384-314 25384-324 25384-326 25384-332 25384-342 25384-344 25384-346 25384-348 89022-320 89038-968 89038-970 89107-632 89137-760 25384-094 25384-316 25384-338 25384-320 25384-328 89230-472 10053-704 10799-140 10799-192 10799-194
25384-062CS 423.63 CAD
25384-062 25384-088 25384-090 25384-092 25384-093 25384-164 25384-166 25384-168 25384-302 25384-304 25384-306 25384-308 25384-310 25384-312 25384-314 25384-324 25384-326 25384-332 25384-342 25384-344 25384-346 25384-348 89022-320 89038-968 89038-970 89107-632 89137-760 25384-094 25384-316 25384-338 25384-320 25384-328 89230-472 10053-704 10799-140 10799-192 10799-194
VWR® Disposable Petri Dishes
Culture Dishes Petri Dishes
Clearly superior, high-quality Petri dishes are manufactured from virgin crystal grade polystyrene for optimum flatness and high optical clarity.
  • Virgin crystal grade polystyrene construction
  • Available in varied sizes and styles for specialized and routine applications
  • Stacking ring and slip-ring features available
  • Supplied sterile
VWR® Petri dishes are available in a variety of sizes and styles to accommodate both specialized and routine needs. The stacking ring feature, on some models, facilitates positive stacking while the slip-ring feature, onMore Product Information