ELIMINASE®, Decontaminants, Decon Labs
Supplier: Decon Labs
1101
10798-804CS 816.88 CAD
10798-804
ELIMINASE®, Decontaminants, Decon Labs
Cleaning Agents Decontaminants
ELIMINase removes RNase, DNase, unwanted DNA and radioactive contamination.
  • Phosphate free and non-toxic
  • Safely disposable and totally rinsable for the decontaminated surface
  • Eliminates the time-consuming method of autoclaving
It works instantly so there is no waiting. ELIMINase is an ideal substitute for using DiEthylPryoCarbonate (DEPC) which is a carcinogen. ELIMINase can be used on all surfaces including glass, plastic, metal and ceramic.