VWR® Basic Refrigerators with Glass Doors and Natural Refrigerant
Supplier: VWR International
10791-618 10791-624 10791-626 10018-860 10018-864 10018-866 75871-648 75871-650
10791-618EA 12272.32 CAD
10791-618 10791-624 10791-626 10018-860 10018-864 10018-866 75871-648 75871-650
VWR® Basic Refrigerators with Glass Doors and Natural Refrigerant
Refrigerators
Swinging or sliding doors. These upright laboratory refrigerators maintain an average temperature of 3 – 5 °C (37 – 41 °F).
  • All units feature a natural refrigerant that is environmentally friendly
  • R290 hydrocarbon refrigerant emits zero ozone depletion potential
  • Units are supplied standard with the LAE temperature control
  • Exterior digital display and alarm function
Units are equipped with low E, double-pane, thermal insulated glass doors for energy efficiency.
Note: Units do not come with castors. Castors can be ordered separately.
Order Now
SPECIFICATIONS
Cat. No. 10791-618 10791-624 10791-626 10018-860 10018-864 10018-866 75871-648 75871-650
Environmentally Preferable Safer Human and Environmental Health,Low Manufacturing Impact,Energy Efficient Safer Human and Environmental Health,Low Manufacturing Impact,Energy Efficient Safer Human and Environmental Health,Low Manufacturing Impact,Energy Efficient Safer Human and Environmental Health,Low Manufacturing Impact,Energy Efficient Safer Human and Environmental Health,Low Manufacturing Impact,Energy Efficient Safer Human and Environmental Health,Low Manufacturing Impact,Energy Efficient
Shelves/drawers 4 full size shelves 8 full size shelves 12 full size shelves 4 full size shelves 8 full size shelves 12 full size shelves 8 full size shelves 8 full size shelves
Door type 1 glass swing door 2 glass swing doors 3 glass swing doors 1 glass swing door 2 glass swing doors 3 glass swing doors 2 glass sliding doors 2 glass sliding doors
Compressor hp ½ hp ½ hp hp ½ hp ½ hp hp ½ hp
Electrical 115 V, 2.4 A 115 V, 5.8 A 115 V, 7.1 A 115 V, 2.4 A 115 V, 5.8 A 115 V, 7.1 A 115 V, 60 Hz, 3.9 A 115 V, 60 Hz, 5.8 A
Weight 161 kg (355 lbs.) 226 kg (497 lbs.) 286 kg (630 lbs.) 161 kg (355 lbs.) 226 kg (497 lbs.) 286 kg (630 lbs.) 197 kg (435 lbs.) 220 kg (485 lbs.)