Human Recombinant PDGF-AB (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-17-100UG 100-17-10UG 100-17-1MG 100-17-500UG
10788-926EA 1001.15 CAD
10788-926 10788-928 10788-930 10788-932 10821-864
Human Recombinant PDGF-AB (from E. coli)
Proteins and Peptides
Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis. PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β and EGF signaling. PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells.
  • High quality
  • Low endotoxin
  • Affordable
PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues. PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB). The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.
Caution: For research use only.