Human Recombinant FGF-Basic 154 (from E. coli)

Supplier: FUJIFILM IRVINE SCIENTIFIC, INC

100-146-50UG 100-146-10UG 100-146-1MG 100-146-500UG 100-146-100UG
10787-332EA 358.96 CAD
10787-332 10788-696 10788-698 10821-764 10821-972
Human Recombinant FGF-Basic 154 (from E. coli)
Proteins and Peptides

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Recombinant FGF-basic 154 is the full length FGF-basic protein encoded by the human FGF-2 gene. FGF-basic 154 is the most popular tissue culture product at Shenandoah Biotechnology, Inc. There are no detectable differences in biological activity between FGF-basic 154 and the truncated FGF-basic 147 proteins.


  • High quality
  • Low endotoxin
  • Affordable


Caution: For research use only.

Order Now

SPECIFICATIONS

Protein/peptide name FGF-Basic 154
Environmentally Preferable
Protein synonyms Prostatropin|HBGF-2|FGF2
Protein/peptide type Recombinant
Protein Function Cytokine
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID P09038
Sequence MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
Protease-free N
Animal-free N
Nuclease-free N
Carrier-free Y
Endotoxin-free N
Endotoxin level Low
Endotoxin content Endotoxin LAL (EU/µg) ≤0.1
Biological activity Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0.5 ng/ml, Bioactivity ≥2.0×10⁶ units/mg
Purity ≥95%
Grade RUO
Formulation 10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7.5
Molecular weight 17.3 kDa
Reconstitution instructions Sterile water at 0.1 mg/ml
Storage conditions –20 °C
Shipping temperature RT
Tested applications Bioassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR