Mouse Recombinant MCP-1 / CCL2 (from E. coli)

Supplier: FUJIFILM IRVINE SCIENTIFIC, INC

200-39-100UG 200-39-10UG 200-39-1MG 200-39-500UG
10788-130EA 884.72 CAD
10788-130 10788-132 10788-134 10788-136 10822-248
Mouse Recombinant MCP-1 / CCL2 (from E. coli)
Proteins and Peptides

Monocyte chemotactic protein 1 (MCP-1), also known as CCL2, is produced by injured or infected tissues. MCP-1 signals through the CCR2 and CCR4 G protein-coupled receptors to recruit memory T cells, monocytes, and dendritic cells to sites of inflammation.


  • High Quality
  • Low Endotoxin
  • Affordable


A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  


Caution: For research use only.

Order Now

SPECIFICATIONS

Protein/peptide name MCP-1 / CCL2 MCP-1 / CCL2
Environmentally Preferable
Protein synonyms JE|MCAF|CCL2 JE|MCAF|CCL2
Protein/peptide type Recombinant Recombinant
Protein Function Cytokine Cytokine
Species Mouse Mouse
Source E. coli E. coli
Conjugation Unconjugated Unconjugated
Gene ID Q5SVU3 Q5SVU3
Sequence QPDAVNAPLT CCYSFTSKMI PMSRLESYKR ITSSRCPKEA VVFVTKLKRE VCADPKKEWV QTYIKNLDRN QMRSEPTTLF KTASALRSSA PLNVKLTRKS EANASTTFST TTSSTSVGVT SVTVN QPDAVNAPLT CCYSFTSKMI PMSRLESYKR ITSSRCPKEA VVFVTKLKRE VCADPKKEWV QTYIKNLDRN QMRSEPTTLF KTASALRSSA PLNVKLTRKS EANASTTFST TTSSTSVGVT SVTVN
Protease-free N N
Animal-free N N
Nuclease-free N N
Carrier-free Y Y
Endotoxin-free N N
Endotoxin level Low Low
Endotoxin content Endotoxin LAL (EU/µg) ≤0.1 Endotoxin LAL (EU/µg) ≤0.1
Biological activity Bioactivity assay: THP-1 chemotaxis, ED50 ≤100 ng/ml, Bioactivity ≥NA Bioactivity assay: THP-1 chemotaxis, ED50 ≤100 ng/ml, Bioactivity ≥NA
Purity ≥95% ≥95%
Grade RUO
Formulation 0.1% Trifluoroacetic Acid (TFA) 0.1% Trifluoroacetic Acid (TFA)
Molecular weight 13.8 kDa 13.8 kDa
Reconstitution instructions Sterile water at 0.1 mg/ml Sterile water at 0.1 mg/ml
Storage conditions –20 °C –20 °C
Shipping temperature RT RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR