Mouse Recombinant IL-3 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-01-100UG 200-01-10UG 200-01-1MG 200-01-500UG
10787-834EA 1154.35 CAD
10787-834 10787-836 10787-838 10787-840 10822-092
Mouse Recombinant IL-3 (from E. coli)
Proteins and Peptides
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.