Human Recombinant IL-22 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-91-100UG 100-91-10UG 100-91-1MG 100-91-500UG
10787-818EA 1368.83 CAD
10787-818 10787-820 10787-822 10787-824 10822-202
Human Recombinant IL-22 (from E. coli)
Proteins and Peptides
Interleukin 22 (IL-22), also called IL-TIF, is an IL-10 family member that is produced by activated dendritic cells and T lymphocytes. IL-22 signals via a heteroduplex receptor consisting of IL-22R and IL-10Rbeta chains. IL-22 is a potent mediator of cellular inflammatory responses. 
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Ordering information: Ships at room temperature; Store at ­20°C upon delivery.