Human Recombinant Omentin (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-140-100UG 100-140-10UG 100-140-1MG 100-140-500UG
10787-290EA 1368.83 CAD
10787-290 10787-292 10787-294 10787-296 10822-322
Human Recombinant Omentin (from E. coli)
Proteins and Peptides
Omentin is an adipokine that is produced and secreted by the small intestine, visceral adipose tissue, perivascular adipose tissue, and epicardial adipose tissue. Omentin enhances insulin-stimulated glucose uptake in adipocytes and is a link between obesity and Type 2 Diabetes. Omentin also functions as a vasodilator and plays a protective role during coronary atherosclerosis and hypertension.
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Ordering information: Ships at room temperature; Store at ­20°C upon delivery.