Human Recombinant EG-VEGF (from E. coli)

Supplier: FUJIFILM IRVINE SCIENTIFIC, INC

100-07-100UG 100-07-1MG 100-07-20UG 100-07-5UG 100-07-500UG
10787-100EA 812.55 CAD
10787-100 10787-102 10787-104 10787-106 10821-930
Human Recombinant EG-VEGF (from E. coli)
Proteins and Peptides

Endocrine gland-derived vascular endothelial growth factor (EG-VEGF) is an angiogenic growth factor that is expressed in the ovaries, testis, adrenal, and placental tissues. EG-VEGF has mitogenic, chemoattractive, and antiapoptotic functional roles.


  • High quality
  • Low endotoxin
  • Affordable


EG-VEGF signaling is mediated through binding the G protein-coupled receptors prokineticin receptor 1 (PKR1) and prokineticin receptor 2 (PKR2).  Polycystic ovaries display strong EG-VEGF expression that is associated with increased angiogenesis and cyst formation, which could lead to the formation of polycystic ovary syndrome and infertility.


Caution: For research use only.

Order Now

SPECIFICATIONS

Protein/peptide name EG-VEGF EG-VEGF
Environmentally Preferable
Protein synonyms Prokineticin 1|PROK1 Prokineticin 1|PROK1
Protein/peptide type Recombinant Recombinant
Protein Function Cytokine Cytokine
Species Human Human
Source E. coli E. coli
Conjugation Unconjugated Unconjugated
Gene ID P58294 P58294
Sequence AVITGACERD VQCGAGTCCA ISLWLRGLRM CTPLGREGEE CHPGSHKVPF FRKRKHHTCP CLPNLLCSRF PDGRYRCSMD LKNINF AVITGACERD VQCGAGTCCA ISLWLRGLRM CTPLGREGEE CHPGSHKVPF FRKRKHHTCP CLPNLLCSRF PDGRYRCSMD LKNINF
Protease-free N N
Animal-free N N
Nuclease-free N N
Carrier-free Y Y
Endotoxin-free N N
Endotoxin level Low Low
Endotoxin content Endotoxin LAL (EU/µg) ≤0.1 Endotoxin LAL (EU/µg) ≤0.1
UniProtKB P58294 P58294
Biological activity Bioactivity: No biological activity data is available at this time Bioactivity: No biological activity data is available at this time
Purity ≥95% ≥95%
Grade RUO RUO
Formulation 0.1% Trifluoroacetic Acid (TFA) 0.1% Trifluoroacetic Acid (TFA)
Molecular weight 9.7 kDa 9.7 kDa
Reconstitution instructions Sterile water at 0.1 mg/ml Sterile water at 0.1 mg/ml
Storage conditions −20 °C −20 °C
Shipping temperature RT RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR