OptiTDS™, Epithelial Tissue Dissociation System, CHI Scientific
Supplier: CHI Scientific
4-20251 4-20252 4-20253 4-20254 4-20255 4-20256 4-20257 4-20258 4-20259 4-20260 4-20261 4-20262 4-20263
10786-126EA 601.28 CAD
10786-126 10786-128 10786-130 10786-132 10786-134 10786-136 10786-138 10786-140 10786-142 10786-144 10786-146 10786-148 10786-150
OptiTDS™, Epithelial Tissue Dissociation System, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.