OptiTDS™, Brain Tissue Dissociation System, CHI Scientific
Supplier: CHI Scientific
4-20221 4-20222 4-20223 4-20224 4-20225 4-20226 4-20227 4-20228
10786-088EA 538.12 CAD
10786-088 10786-090 10786-092 10786-094 10786-096 10786-098 10786-100 10786-102
OptiTDS™, Brain Tissue Dissociation System, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.